Assay Details
Target Gene Details
Entrez Gene ID: | 57205 |
Gene Name: | ATPase phospholipid transporting 10D (putative) |
Gene Aliases: |
ATPVD |
Location: |
Chr.4:47485261-47593486 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP10D | NM_020453.3 | NP_065186.3 | ||
| XM_005248119.4 | 1 | 149 | XP_005248176.1 | |
| XM_005248120.4 | 1 | 136 | XP_005248177.1 | |
| XM_011513722.2 | 1 | 149 | XP_011512024.1 | |
| XM_017008472.1 | 1 | 129 | XP_016863961.1 | |
| AJ441078.1 | CAD29577.1 | |||
| BC057839.1 | 1 | 112 | AAH57839.1 | |
| BC131535.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829923 | Chr.4:47434122 - 47676426 on Build GRCh38 | Gain |
|
| esv2727527 | Chr.4:47485160 - 47486339 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map