Assay Details
Target Gene Details
Entrez Gene ID: | 84705 |
Gene Name: | GTP binding protein 3 (mitochondrial) |
Gene Aliases: |
COXPD23, GTPBG3, MSS1, MTGP1, THDF1 |
Location: |
Chr.19:17334982-17342731 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GTPBP3 | NM_001128855.2 | 7 | 993 | NP_001122327.1 |
| NM_001195422.1 | 7 | 986 | NP_001182351.1 | |
| NM_032620.3 | 7 | 993 | NP_116009.2 | |
| NM_133644.3 | 6 | 1089 | NP_598399.2 | |
| AF360742.1 | 7 | 955 | AAK39555.1 | |
| AF360743.1 | 7 | 955 | AAK39556.1 | |
| AF360744.1 | 7 | 935 | AAK39557.1 | |
| AK027606.1 | 7 | 940 | BAB55228.1 | |
| AK091035.1 | 6 | 1794 | ||
| AK291929.1 | 7 | 937 | ||
| AK297953.1 | 7 | 968 | ||
| AK310794.1 | ||||
| AK311442.1 | 7 | 1056 | ||
| AY078987.1 | 6 | 1051 | AAL85492.1 | |
| AY078988.1 | 7 | 955 | AAL85493.1 | |
| BC008924.2 | 5 | 533 | ||
| BC017207.2 | 7 | 925 | AAH17207.1 | |
| BC019261.1 | 7 | 934 | AAH19261.1 | |
| BM789389.1 | 4 | 457 | ||
| DA490131.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833768 | Chr.19:17338840 - 17526738 on Build GRCh38 | Loss |
|
| nsv578741 | Chr.19:17335702 - 17339637 on Build GRCh38 | Gain |
|
| nsv470129 | Chr.19:17188084 - 17341718 on Build GRCh38 | Loss |
|
| esv21604 | Chr.19:17332440 - 17339470 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map