Assay Details
Target Gene Details
Entrez Gene ID: | 7988 |
Gene Name: | zinc finger protein 212 |
Gene Aliases: |
C2H2-150, ZNF182, ZNFC150 |
Location: |
Chr.7:149239648-149255609 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF212 | NM_012256.3 | 5 | 1073 | NP_036388.2 |
| XM_005250055.2 | 5 | 1071 | XP_005250112.2 | |
| XM_011516581.2 | 5 | 1058 | XP_011514883.2 | |
| XM_011516582.1 | 5 | 993 | XP_011514884.1 | |
| XM_017012646.1 | 5 | 1071 | XP_016868135.1 | |
| XM_017012647.1 | 6 | 1027 | XP_016868136.1 | |
| XM_017012648.1 | 5 | 1316 | XP_016868137.1 | |
| AK074821.1 | 6 | 1076 | ||
| AK315086.1 | 5 | 1082 | ||
| BC022785.2 | 5 | 1040 | AAH22785.2 | |
| U38864.1 | 3 | 523 | AAC51296.1 |
More Information
Set Membership: |
|

Genomic Map