Assay Details
Target Gene Details
Entrez Gene ID: | 23205 |
Gene Name: | acyl-CoA synthetase bubblegum family member 1 |
Gene Aliases: |
BG, BG1, BGM, GR-LACS, LPD |
Location: |
Chr.15:78167468-78234707 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACSBG1 | NM_001199377.1 | 9 | 1362 | NP_001186306.1 |
| NM_015162.4 | 9 | 1374 | NP_055977.3 | |
| XM_011521390.2 | 9 | 1188 | XP_011519692.1 | |
| XM_011521391.2 | 8 | 3047 | XP_011519693.2 | |
| XM_017022025.1 | 9 | 1208 | XP_016877514.1 | |
| XM_017022026.1 | 12 | 2009 | XP_016877515.1 | |
| AF179481.1 | 9 | 1215 | AAG09404.1 | |
| AK054882.1 | 9 | 1224 | ||
| AK294202.1 | 5 | 660 | ||
| AK295269.1 | 9 | 1180 | ||
| AK299597.1 | 8 | 1140 | ||
| AK314508.1 | 9 | 1374 | ||
| BC009289.2 | 9 | 1173 | AAH09289.1 |
More Information
Set Membership: |
|

Genomic Map