Assay Details
Target Gene Details
Entrez Gene ID: | 261734 |
Gene Name: | nephronophthisis 4 |
Gene Aliases: |
POC10, SLSN4 |
Location: |
Chr.1:5862808-5992473 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NPHP4 | NM_001291593.1 | 17 | 2707 | NP_001278522.1 |
| NM_001291594.1 | 16 | 2530 | NP_001278523.1 | |
| NM_015102.4 | 20 | 3017 | NP_055917.1 | |
| NR_111987.1 | 21 | 3301 | ||
| XM_006710563.3 | 20 | 2863 | XP_006710626.1 | |
| XM_011541213.1 | 19 | 2784 | XP_011539515.1 | |
| XM_011541214.1 | 19 | 2787 | XP_011539516.1 | |
| XM_011541215.1 | 18 | 2676 | XP_011539517.1 | |
| XM_011541216.2 | 21 | 3491 | XP_011539518.1 | |
| XM_011541217.2 | 20 | 3355 | XP_011539519.1 | |
| XM_011541218.2 | 21 | 3024 | XP_011539520.1 | |
| XM_017000996.1 | 19 | 2784 | XP_016856485.1 | |
| XM_017000997.1 | 20 | 3286 | XP_016856486.1 | |
| XM_017000998.1 | 19 | 2787 | XP_016856487.1 | |
| XM_017000999.1 | 20 | 2782 | XP_016856488.1 | |
| XM_017001000.1 | 19 | 2615 | XP_016856489.1 | |
| XM_017001001.1 | 14 | 2054 | XP_016856490.1 | |
| XM_017001002.1 | 19 | 2787 | XP_016856491.1 | |
| XM_017001003.1 | 12 | 1845 | XP_016856492.1 | |
| AB014573.1 | 15 | 2117 | BAA31648.1 | |
| AF537130.1 | 20 | 3015 | AAN06814.1 | |
| AK124074.1 | 16 | 2483 | ||
| AK128182.1 | 2 | 2874 | ||
| AK314326.1 | 17 | 2681 | ||
| AY118228.1 | 20 | 2952 | AAM78558.1 | |
| BC040520.1 | 21 | 3302 | AAH40520.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv545173 | Chr.1:5833391 - 6025293 on Build GRCh38 | Gain |
|
| nsv508723 | Chr.1:5777215 - 5971948 on Build GRCh38 | Insertion |
|
| nsv545174 | Chr.1:5844658 - 5971884 on Build GRCh38 | Loss |
|
| nsv831203 | Chr.1:5830030 - 5967902 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114545322] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map