Assay Details
Target Gene Details
Entrez Gene ID: | 402 |
Gene Name: | ADP ribosylation factor like GTPase 2 |
Gene Aliases: |
ARFL2 |
Location: |
Chr.11:65014113-65022185 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARL2 | NM_001199745.1 | NP_001186674.1 | ||
| NM_001667.3 | NP_001658.2 | |||
| AF493888.1 | AAM12602.1 | |||
| AK223502.1 | BAD97222.1 | |||
| AK294580.1 | ||||
| BC002530.2 | AAH02530.1 | |||
| BG706294.1 | ||||
| BT006674.1 | AAP35320.1 | |||
| CN338497.1 | ||||
| L13687.1 | AAC37606.1 |
Target Gene Details
Entrez Gene ID: | 100528018 |
Gene Name: | ARL2-SNX15 readthrough (NMD candidate) |
Gene Aliases: |
- |
Location: |
Chr.11:65014113-65040572 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARL2-SNX15 | NR_037650.1 | |||
| AK096447.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526181 | Chr.11:64870569 - 65489858 on Build GRCh38 | Gain |
|
| esv3626667 | Chr.11:65009442 - 65019708 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map