Assay Details
Target Gene Details
Entrez Gene ID: | 85440 |
Gene Name: | dedicator of cytokinesis 7 |
Gene Aliases: |
EIEE23, ZIR2 |
Location: |
Chr.1:62454726-62688368 on Build GRCh38 |
Assay Gene Location: | Within Exon 51 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DOCK7 | NM_001271999.1 | 47 | 6225 | NP_001258928.1 |
| NM_001272000.1 | 47 | 6138 | NP_001258929.1 | |
| NM_001272001.1 | 46 | 6132 | NP_001258930.1 | |
| NM_033407.3 | 47 | 6165 | NP_212132.2 | |
| XM_005271292.2 | 48 | 6221 | XP_005271349.1 | |
| XM_011542326.2 | 48 | 6248 | XP_011540628.1 | |
| XM_011542327.2 | 47 | 6242 | XP_011540629.1 | |
| XM_011542328.2 | 47 | 6233 | XP_011540630.1 | |
| XM_017002639.1 | 46 | 6149 | XP_016858128.1 | |
| AB051558.1 | 28 | 3641 | BAB21862.1 | |
| AK055493.1 | 14 | 1782 | BAB70933.1 | |
| AK055905.1 | 19 | 2657 | BAB71042.1 | |
| AK091479.1 | 21 | 2632 | ||
| AK125049.1 | 33 | 4310 | BAC86032.1 | |
| AK299737.1 | 19 | 2348 | ||
| BC016392.1 | 16 | 2129 | ||
| BC112387.1 | 7 | 884 | ||
| DQ118679.1 | 44 | 5710 | AAZ38451.1 | |
| DQ118680.1 | 43 | 5704 | AAZ38452.1 | |
| DQ341187.1 | 47 | 6132 |
More Information
Set Membership: |
|

Genomic Map