Assay Details
Target Gene Details
Entrez Gene ID: | 9001 |
Gene Name: | huntingtin associated protein 1 |
Gene Aliases: |
HAP2, HIP5, HLP, hHLP1 |
Location: |
Chr.17:41722639-41734646 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HAP1 | NM_001079870.1 | 10 | 2782 | NP_001073339.1 |
| NM_001079871.1 | 10 | 2758 | NP_001073340.1 | |
| NM_177977.2 | 11 | 2833 | NP_817084.2 | |
| AB209105.1 | 10 | 2743 | BAD92342.1 | |
| AF040723.1 | 11 | 2831 | AAC39861.1 | |
| AK022007.1 | 10 | 2779 | BAB13952.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758690 | Chr.17:41145123 - 41740591 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map