Assay Details
Target Gene Details
Entrez Gene ID: | 149420 |
Gene Name: | PDLIM1 interacting kinase 1 like |
Gene Aliases: |
CLIK1L, STK35L2 |
Location: |
Chr.1:26109406-26125548 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDIK1L | NM_001243532.1 | 3 | 1670 | NP_001230461.1 |
| NM_001243533.1 | 4 | 1744 | NP_001230462.1 | |
| NM_152835.4 | 3 | 1553 | NP_690048.1 | |
| XM_005245743.4 | 4 | 1813 | XP_005245800.1 | |
| XM_006710381.3 | 2 | 3127 | XP_006710444.1 | |
| XM_011540792.2 | 2 | 1224 | XP_011539094.1 | |
| AF411102.1 | 4 | 1684 | AAN03661.1 | |
| AL834290.1 | 1 | 999 | CAD38964.1 | |
| BC028713.1 | 4 | 1686 | AAH28713.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv950339 | Chr.1:26121910 - 26127709 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map