Assay Details
Target Gene Details
Entrez Gene ID: | 84002 |
Gene Name: | UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 5 |
Gene Aliases: |
B3GN-T5, beta3Gn-T5 |
Location: |
Chr.3:183253207-183273391 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| B3GNT5 | NM_032047.4 | 2 | 489 | NP_114436.1 |
| XM_005247823.4 | 2 | 694 | XP_005247880.1 | |
| XM_005247824.3 | 3 | 748 | XP_005247881.1 | |
| XM_005247825.3 | 3 | 632 | XP_005247882.1 | |
| XM_011513225.2 | 2 | 1128 | XP_011511527.1 | |
| XM_011513226.2 | 3 | 605 | XP_011511528.1 | |
| XM_011513227.2 | 3 | 639 | XP_011511529.1 | |
| XM_011513228.2 | 2 | 540 | XP_011511530.1 | |
| XM_011513229.2 | 2 | 394 | XP_011511531.1 | |
| XM_017007309.1 | 3 | 1762 | XP_016862798.1 | |
| AB045278.1 | 1 | 88 | BAB40940.1 | |
| AB209517.1 | 2 | 307 | BAD92754.1 | |
| AF368169.1 | 1 | 422 | AAK53403.1 | |
| AJ304505.1 | CAC83093.1 | |||
| AK074424.1 | 2 | 476 | ||
| AK091474.1 | 2 | 676 | ||
| BP270576.1 | 2 | 489 |
Target Gene Details
Entrez Gene ID: | 23101 |
Gene Name: | MCF.2 cell line derived transforming sequence-like 2 |
Gene Aliases: |
ARHGEF22 |
Location: |
Chr.3:183176116-183428990 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MCF2L2 | NM_015078.3 | NP_055893.3 | ||
| XM_011512585.2 | XP_011510887.1 | |||
| XM_011512586.2 | XP_011510888.1 | |||
| XM_017005943.1 | XP_016861432.1 | |||
| XM_017005945.1 | XP_016861434.1 | |||
| XM_017005946.1 | XP_016861435.1 | |||
| AB020668.1 | BAA74884.1 | |||
| AK301698.1 | ||||
| AY172737.1 | AAO19651.1 | |||
| BC064632.1 | AAH64632.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893833 | Chr.3:183130677 - 183342208 on Build GRCh38 | Gain |
|
| nsv1006872 | Chr.3:183194738 - 183548501 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map