Assay Details
Target Gene Details
Entrez Gene ID: | 10691 |
Gene Name: | glucocorticoid modulatory element binding protein 1 |
Gene Aliases: |
P96PIF, PIF96 |
Location: |
Chr.1:28668728-28715603 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GMEB1 | NM_001319674.1 | NP_001306603.1 | ||
| NM_006582.3 | NP_006573.2 | |||
| NM_024482.2 | NP_077808.1 | |||
| XM_011540518.2 | XP_011538820.1 | |||
| XM_011540519.2 | XP_011538821.1 | |||
| XM_011540521.2 | XP_011538823.1 | |||
| XM_011540522.2 | XP_011538824.1 | |||
| XM_017000087.1 | XP_016855576.1 | |||
| AF099013.1 | AAD39355.1 | |||
| AF173868.1 | AAD51352.1 | |||
| AF203694.1 | AAG01189.1 | |||
| AK000892.1 | BAA91410.1 | |||
| AK023126.1 | ||||
| AK315195.1 | ||||
| AK315436.1 | ||||
| BC001473.2 | AAH01473.1 | |||
| DA647317.1 | ||||
| DA919003.1 |
More Information
Set Membership: |
|

Genomic Map