Assay Details
Target Gene Details
Entrez Gene ID: | 65108 |
Gene Name: | MARCKS like 1 |
Gene Aliases: |
F52, MACMARCKS, MLP, MLP1, MRP |
Location: |
Chr.1:32333829-32336239 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MARCKSL1 | NM_023009.6 | NP_075385.1 | ||
| NR_052852.1 | ||||
| AI879076.1 | ||||
| AL713653.1 | CAD28462.1 | |||
| AY007131.1 | ||||
| BC007904.2 | AAH07904.1 | |||
| BC066915.1 | AAH66915.1 | |||
| DT218825.1 | ||||
| X70326.1 | CAA49793.1 |
More Information
Set Membership: |
|

Genomic Map