Assay Details
Target Gene Details
Entrez Gene ID: | 8424 |
Gene Name: | gamma-butyrobetaine hydroxylase 1 |
Gene Aliases: |
BBH, BBOX, G-BBH, gamma-BBH |
Location: |
Chr.11:27040705-27127807 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BBOX1 | NM_003986.2 | 4 | 631 | NP_003977.1 |
| XM_005253159.2 | 4 | 879 | XP_005253216.1 | |
| XM_005253160.2 | 4 | 876 | XP_005253217.1 | |
| XM_005253161.2 | 4 | 628 | XP_005253218.1 | |
| XM_011520402.1 | 5 | 1039 | XP_011518704.1 | |
| XM_017018403.1 | 3 | 561 | XP_016873892.1 | |
| AF082868.1 | 3 | 329 | AAC64066.1 | |
| AK290108.1 | 3 | 376 | ||
| AK313422.1 | 3 | 379 | ||
| BC011034.1 | 2 | 319 | AAH11034.1 | |
| CR456812.1 | 2 | 263 | CAG33093.1 |
Target Gene Details
Entrez Gene ID: | 103695435 |
Gene Name: | BBOX1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.11:27047186-27220113 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BBOX1-AS1 | NR_125768.1 | |||
| DA865512.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1727n54 | Chr.11:26992485 - 27219404 on Build GRCh38 | Gain |
|
| esv3625759 | Chr.11:26962165 - 27229739 on Build GRCh38 | Gain |
|
| nsv7720 | Chr.11:27050452 - 27059150 on Build GRCh38 | Deletion |
|
| nsv508619 | Chr.11:27006947 - 27089187 on Build GRCh38 | Deletion |
|
| esv3625760 | Chr.11:26964078 - 27108838 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map