Assay Details
Target Gene Details
Entrez Gene ID: | 257101 |
Gene Name: | zinc finger protein 683 |
Gene Aliases: |
Hobit |
Location: |
Chr.1:26361632-26374520 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF683 | NM_001114759.2 | 6 | 1374 | NP_001108231.1 |
| NM_001307925.1 | 7 | 1431 | NP_001294854.1 | |
| NM_173574.3 | 6 | 1371 | NP_775845.2 | |
| XM_005245828.3 | 7 | 1548 | XP_005245885.1 | |
| XM_005245830.3 | 7 | 1429 | XP_005245887.1 | |
| XM_005245832.3 | 7 | 1454 | XP_005245889.1 | |
| XM_006710555.3 | 8 | 1632 | XP_006710618.1 | |
| XM_011541198.2 | 7 | 1451 | XP_011539500.1 | |
| XM_017000954.1 | 7 | 1551 | XP_016856443.1 | |
| XM_017000955.1 | 7 | 1429 | XP_016856444.1 | |
| XM_017000956.1 | 7 | 1426 | XP_016856445.1 | |
| XM_017000957.1 | 6 | 1491 | XP_016856446.1 | |
| XM_017000958.1 | 7 | 1531 | XP_016856447.1 | |
| XM_017000959.1 | 6 | 1471 | XP_016856448.1 | |
| BC028731.2 | 6 | 1315 | AAH28731.2 | |
| BC029505.1 | 7 | 1412 | AAH29505.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv545907 | Chr.1:26352550 - 26370135 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map