Assay Details
Target Gene Details
Entrez Gene ID: | 374864 |
Gene Name: | coiled-coil domain containing 178 |
Gene Aliases: |
C18orf34 |
Location: |
Chr.18:32937402-33441040 on Build GRCh38 |
Assay Gene Location: | Within Exon 29 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC178 | NM_001105528.1 | 22 | 3119 | NP_001098998.1 |
| NM_001308126.1 | 23 | 3191 | NP_001295055.1 | |
| NM_198995.2 | 22 | 3074 | NP_945346.2 | |
| XM_011525948.1 | 23 | 3188 | XP_011524250.1 | |
| XM_011525951.1 | 22 | 3038 | XP_011524253.1 | |
| XM_017025721.1 | 24 | 3260 | XP_016881210.1 | |
| XM_017025722.1 | 24 | 3281 | XP_016881211.1 | |
| XM_017025723.1 | 24 | 3290 | XP_016881212.1 | |
| XM_017025724.1 | 18 | 2824 | XP_016881213.1 | |
| XM_017025725.1 | 18 | 3441 | XP_016881214.1 | |
| AK000027.1 | 3 | 640 | ||
| AK126038.1 | BAC86407.1 | |||
| AK129955.1 | 9 | 1468 | BAC85257.1 | |
| BC130535.1 | ||||
| BC144178.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758720 | Chr.18:32803364 - 32994773 on Build GRCh38 | Loss |
|
| dgv3333n100 | Chr.18:32901167 - 32960383 on Build GRCh38 | Loss |
|
| nsv528397 | Chr.18:32918657 - 32977021 on Build GRCh38 | Loss |
|

Genomic Map