Assay Details
Target Gene Details
Entrez Gene ID: | 256949 |
Gene Name: | KN motif and ankyrin repeat domains 3 |
Gene Aliases: |
ANKRD47 |
Location: |
Chr.19:8322584-8343262 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KANK3 | NM_198471.2 | NP_940873.2 | ||
| XM_006722718.3 | XP_006722781.1 | |||
| XM_011527884.2 | XP_011526186.1 | |||
| XM_017026563.1 | XP_016882052.1 | |||
| AB284126.1 | ||||
| AK128815.1 | BAC87620.1 | |||
| BC066124.1 | AAH66124.1 |
More Information
Set Membership: |
|

Genomic Map