Assay Details
Target Gene Details
Entrez Gene ID: | 158800 |
Gene Name: | Rhox homeobox family member 1 |
Gene Aliases: |
OTEX, PEPP1 |
Location: |
Chr.X:120109049-120117651 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RHOXF1 | NM_139282.2 | 1 | 242 | NP_644811.1 |
| XM_011531281.2 | 2 | 293 | XP_011529583.1 | |
| AY099086.1 | 1 | 242 | AAM22794.1 | |
| BC069324.1 | 1 | 217 | AAH69324.1 | |
| BC069529.1 | 1 | 218 | AAH69529.1 | |
| BC103857.1 | 1 | 201 | AAI03858.1 | |
| BC103858.1 | 1 | 201 | AAI03859.1 | |
| BE675048.1 | 3 | 539 |
Target Gene Details
Entrez Gene ID: | 101928969 |
Gene Name: | RHOXF1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.X:120036236-120146855 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RHOXF1-AS1 | NR_131238.1 | |||
| AK123976.1 | ||||
| DA929866.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526653 | Chr.X:120029666 - 120140727 on Build GRCh38 | Gain |
|
| esv2758888 | Chr.X:119823476 - 120195131 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map