Assay Details
Target Gene Details
Entrez Gene ID: | 6730 |
Gene Name: | signal recognition particle 68 |
Gene Aliases: |
- |
Location: |
Chr.17:76038775-76072526 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SRP68 | NM_001260502.1 | NP_001247431.1 | ||
| NM_001260503.1 | NP_001247432.1 | |||
| NM_014230.3 | NP_055045.2 | |||
| NR_048541.1 | ||||
| AF195951.1 | AAF24308.1 | |||
| AK002002.1 | ||||
| AK027320.1 | BAB55040.1 | |||
| AK074698.1 | BAC11145.1 | |||
| AK092935.1 | ||||
| AK097962.1 | ||||
| AK126258.1 | ||||
| AK301100.1 | ||||
| AL512708.1 | ||||
| BC020238.1 | AAH20238.1 | |||
| DQ786315.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833546 | Chr.17:76003077 - 76186972 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map