Assay Details
Target Gene Details
Entrez Gene ID: | 669 |
Gene Name: | bisphosphoglycerate mutase |
Gene Aliases: |
DPGM |
Location: |
Chr.7:134646779-134679816 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BPGM | NM_001293085.1 | 3 | 861 | NP_001280014.1 |
| NM_001724.4 | 2 | 736 | NP_001715.1 | |
| NM_199186.2 | 3 | 1057 | NP_954655.1 | |
| XM_011516527.1 | 2 | 917 | XP_011514829.1 | |
| AK097778.1 | 2 | 680 | ||
| AK315439.1 | 2 | 704 | ||
| BC017050.1 | 2 | 707 | AAH17050.1 | |
| X04327.1 | 2 | 626 | CAA27858.1 |
More Information
Set Membership: |
|

Genomic Map