Assay Details
Target Gene Details
Entrez Gene ID: | 51422 |
Gene Name: | protein kinase AMP-activated non-catalytic subunit gamma 2 |
Gene Aliases: |
AAKG, AAKG2, CMH6, H91620p, WPWS |
Location: |
Chr.7:151556114-151877231 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRKAG2 | NM_016203.3 | 1 | 154 | NP_057287.2 |
| XM_005250002.3 | XP_005250059.1 | |||
| AI084919.1 | 1 | 154 | ||
| AK074675.1 | 1 | 48 | BAC11129.1 | |
| DA123009.1 | 1 | 81 |
Target Gene Details
Entrez Gene ID: | 100505483 |
Gene Name: | PRKAG2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.7:151877042-151879223 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRKAG2-AS1 | NR_038926.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1027917 | Chr.7:151791285 - 151893200 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map