Assay Details
Target Gene Details
Entrez Gene ID: | 3625 |
Gene Name: | inhibin beta B subunit |
Gene Aliases: |
- |
Location: |
Chr.2:120346143-120351807 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| INHBB | NM_002193.2 | 2 | 1107 | NP_002184.2 |
| BC030029.2 | 2 | 1107 | AAH30029.1 | |
| M13437.1 | 2 | 901 | AAA59169.1 | |
| M31682.1 | 2 | 1000 | AAA59170.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv954803 | Chr.2:120343425 - 120390624 on Build GRCh38 | Deletion |
|
| nsv582787 | Chr.2:120343260 - 120352811 on Build GRCh38 | Loss |
|
| nsv582786 | Chr.2:120338344 - 120352811 on Build GRCh38 | Loss |
|
| nsv829640 | Chr.2:120324276 - 120378304 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map