Assay Details
Target Gene Details
Entrez Gene ID: | 9032 |
Gene Name: | transmembrane 4 L six family member 5 |
Gene Aliases: |
- |
Location: |
Chr.17:4771892-4783211 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TM4SF5 | NM_003963.2 | 4 | 549 | NP_003954.2 |
| AF027204.1 | 4 | 550 | AAB82947.1 | |
| BC069519.1 | 4 | 549 | AAH69519.1 | |
| BC093688.1 | 4 | 546 | AAH93688.1 | |
| BC117163.1 | 4 | 546 | ||
| CR456925.1 | 4 | 518 | CAG33206.1 | |
| CR541798.1 | 4 | 518 | CAG46597.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv574273 | Chr.17:4585589 - 5460332 on Build GRCh38 | Gain |
|
| nsv833346 | Chr.17:4646256 - 4860871 on Build GRCh38 | Loss |
|

Genomic Map