Assay Details
Target Gene Details
Entrez Gene ID: | 54619 |
Gene Name: | cyclin J |
Gene Aliases: |
bA690P14.1 |
Location: |
Chr.10:96042848-96060870 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCNJ | NM_001134375.1 | NP_001127847.1 | ||
| NM_001134376.1 | NP_001127848.1 | |||
| NM_019084.4 | NP_061957.2 | |||
| XM_005269921.3 | XP_005269978.1 | |||
| XM_011539885.2 | XP_011538187.1 | |||
| XM_017016354.1 | XP_016871843.1 | |||
| XM_017016355.1 | XP_016871844.1 | |||
| XM_017016356.1 | XP_016871845.1 | |||
| AK001757.1 | BAA91887.1 | |||
| AK092360.1 | ||||
| AK297264.1 | ||||
| AK303018.1 | ||||
| BC043175.1 | AAH43175.1 | |||
| BM789816.1 |
Target Gene Details
Entrez Gene ID: | 728558 |
Gene Name: | ENTPD1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.10:95753206-96090235 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ENTPD1-AS1 | NR_038444.1 | |||
| NR_134320.1 | ||||
| NR_134321.1 | ||||
| NR_134322.1 | ||||
| AI571323.1 | ||||
| AK091396.1 | ||||
| AK096469.1 | ||||
| AL120918.1 | ||||
| BU852745.1 | ||||
| DA419635.1 |
More Information
Set Membership: |
|

Genomic Map