Assay Details
Target Gene Details
Entrez Gene ID: | 1176 |
Gene Name: | adaptor related protein complex 3 sigma 1 subunit |
Gene Aliases: |
CLAPS3, Sigma3A |
Location: |
Chr.5:115841606-115914081 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AP3S1 | NM_001002924.2 | NP_001002924.2 | ||
| NM_001284.3 | NP_001275.1 | |||
| NM_001318090.1 | NP_001305019.1 | |||
| NM_001318091.1 | NP_001305020.1 | |||
| NM_001318093.1 | NP_001305022.1 | |||
| NM_001318094.1 | NP_001305023.1 | |||
| XM_011543136.2 | XP_011541438.2 | |||
| XM_017009024.1 | XP_016864513.1 | |||
| XM_017009025.1 | XP_016864514.1 | |||
| BP368651.1 |
Target Gene Details
Entrez Gene ID: | 9140 |
Gene Name: | autophagy related 12 |
Gene Aliases: |
APG12, APG12L, FBR93, HAPG12 |
Location: |
Chr.5:115828196-115841851 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATG12 | NM_001277783.1 | 1 | 228 | NP_001264712.1 |
| NM_004707.3 | 1 | 228 | NP_004698.3 | |
| NR_033362.1 | 1 | 228 | ||
| NR_033363.1 | 1 | 228 | ||
| NR_073603.1 | 1 | 228 | ||
| AK313320.1 | 1 | 223 | ||
| BC012266.2 | 1 | 179 | AAH12266.2 | |
| BP335591.1 | 1 | 228 | ||
| DA005347.1 | 1 | 224 | ||
| EU554057.1 | 1 | 70 |
More Information
Set Membership: |
|

Genomic Map