Assay Details
Target Gene Details
Entrez Gene ID: | 3026 |
Gene Name: | hyaluronan binding protein 2 |
Gene Aliases: |
FSAP, HABP, HGFAL, NMTC5, PHBP |
Location: |
Chr.10:113550831-113589602 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HABP2 | NM_001177660.2 | NP_001171131.1 | ||
| NM_004132.4 | NP_004123.1 | |||
| AK290832.1 | ||||
| AK303759.1 | ||||
| AK303761.1 | ||||
| AK303948.1 | ||||
| BC031412.1 | AAH31412.1 | |||
| D49742.1 | BAA08576.1 | |||
| S83182.1 | AAB46909.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1048819 | Chr.10:113499347 - 113757619 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map