Assay Details
Target Gene Details
Entrez Gene ID: | 8820 |
Gene Name: | HESX homeobox 1 |
Gene Aliases: |
ANF, CPHD5, RPX |
Location: |
Chr.3:57197838-57227643 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HESX1 | NM_003865.2 | 2 | 610 | NP_003856.1 |
| XM_005265526.4 | 3 | 1734 | XP_005265583.1 | |
| XM_006713379.3 | 3 | 504 | XP_006713442.1 | |
| XM_011534204.2 | 4 | 1259 | XP_011532506.1 | |
| XM_011534205.2 | 4 | 530 | XP_011532507.1 | |
| XM_017007421.1 | 4 | 594 | XP_016862910.1 | |
| AI652412.1 | ||||
| BC069515.1 | 2 | 345 | AAH69515.1 | |
| BC093979.1 | 2 | 322 | AAH93979.1 | |
| BC112089.1 | 2 | 322 | AAI12090.1 |
More Information
Set Membership: |
|

Genomic Map