Assay Details
Target Gene Details
Entrez Gene ID: | 51763 |
Gene Name: | inositol polyphosphate-5-phosphatase K |
Gene Aliases: |
PPS, SKIP |
Location: |
Chr.17:1494575-1516888 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| INPP5K | NM_001135642.1 | NP_001129114.1 | ||
| NM_016532.3 | NP_057616.2 | |||
| NM_130766.2 | NP_570122.1 | |||
| XM_005256683.2 | XP_005256740.1 | |||
| XM_005256685.1 | XP_005256742.1 | |||
| XM_005256686.1 | XP_005256743.1 | |||
| XM_011523934.1 | XP_011522236.1 | |||
| XM_011523936.2 | XP_011522238.1 | |||
| XM_017024755.1 | XP_016880244.1 | |||
| XM_017024756.1 | XP_016880245.1 | |||
| XM_017024757.1 | XP_016880246.1 | |||
| XM_017024758.1 | XP_016880247.1 | |||
| XM_017024759.1 | XP_016880248.1 | |||
| AB036829.1 | BAA92340.1 | |||
| AB036830.1 | BAA92341.1 | |||
| AK297955.1 | ||||
| AK303907.1 | ||||
| AK312585.1 | ||||
| AK312844.1 | ||||
| BC004362.1 | AAH04362.1 | |||
| U45973.1 | AAB03214.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833334 | Chr.17:1411327 - 1582381 on Build GRCh38 | Loss |
|
| esv3582474 | Chr.17:1494346 - 1505407 on Build GRCh38 | Loss |
|
| nsv952098 | Chr.17:1464507 - 1515106 on Build GRCh38 | Deletion |
|
| nsv1057782 | Chr.17:1351585 - 1512616 on Build GRCh38 | Gain |
|
| dgv877e212 | Chr.17:1494279 - 1500654 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map