Assay Details
Target Gene Details
Entrez Gene ID: | 7139 |
Gene Name: | troponin T2, cardiac type |
Gene Aliases: |
CMD1D, CMH2, CMPD2, LVNC6, RCM3, TnTC, cTnT |
Location: |
Chr.1:201359008-201377828 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TNNT2 | NM_000364.3 | 16 | 1151 | NP_000355.2 |
| NM_001001430.2 | 16 | 1130 | NP_001001430.1 | |
| NM_001001431.2 | 15 | 1121 | NP_001001431.1 | |
| NM_001001432.2 | 15 | 1112 | NP_001001432.1 | |
| NM_001276345.1 | 17 | 1160 | NP_001263274.1 | |
| NM_001276346.1 | 15 | 1031 | NP_001263275.1 | |
| NM_001276347.1 | 16 | 1263 | NP_001263276.1 | |
| XM_006711508.3 | XP_006711571.1 | |||
| XM_006711509.3 | XP_006711572.1 | |||
| XM_011509938.2 | XP_011508240.1 | |||
| XM_011509939.1 | XP_011508241.1 | |||
| XM_011509940.2 | XP_011508242.1 | |||
| XM_011509941.2 | XP_011508243.1 | |||
| XM_011509942.2 | XP_011508244.1 | |||
| XM_011509943.2 | XP_011508245.1 | |||
| XM_011509944.2 | XP_011508246.1 | |||
| XM_011509946.1 | XP_011508248.1 | |||
| XM_017002216.1 | XP_016857705.1 | |||
| XM_017002217.1 | XP_016857706.1 | |||
| AA865519.1 | 1 | 74 | ||
| AK055533.1 | ||||
| AK125236.1 | ||||
| AK290621.1 | ||||
| AL832707.1 | ||||
| AY277394.1 | AAP96757.1 | |||
| BC002653.2 | AAH02653.1 | |||
| L40162.1 | AAA67422.1 | |||
| S64668.1 | AAB27731.1 | |||
| X74819.1 | CAA52818.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470776 | Chr.1:201319827 - 201534604 on Build GRCh38 | Gain |
|
| nsv467961 | Chr.1:201316584 - 201534604 on Build GRCh38 | Gain |
|
| nsv947466 | Chr.1:201308515 - 201540498 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map