Assay Details
Target Gene Details
Entrez Gene ID: | 7262 |
Gene Name: | pleckstrin homology like domain family A member 2 |
Gene Aliases: |
BRW1C, BWR1C, HLDA2, IPL, TSSC3 |
Location: |
Chr.11:2928273-2929420 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PHLDA2 | NM_003311.3 | 2 | 733 | NP_003302.1 |
| AF001294.1 | AAB86680.1 | |||
| AF019953.1 | AAC51912.1 | |||
| AF035444.1 | 2 | 733 | AAC17458.1 | |
| AI659266.1 | ||||
| AK223027.1 | 2 | 733 | BAD96747.1 | |
| BC005034.1 | 2 | 723 | AAH05034.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759795 | Chr.11:2767814 - 2942382 on Build GRCh38 | Gain |
|
| nsv832050 | Chr.11:2868540 - 2963987 on Build GRCh38 | Loss |
|
| nsv553121 | Chr.11:2917730 - 2928631 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map