Assay Details
Target Gene Details
Entrez Gene ID: | 346689 |
Gene Name: | killer cell lectin like receptor G2 |
Gene Aliases: |
CLEC15B |
Location: |
Chr.7:139426931-139483716 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLRG2 | NM_198508.2 | 1 | 198 | NP_940910.1 |
| XM_005250311.3 | 1 | 203 | XP_005250368.1 | |
| XM_011516140.2 | 1 | 203 | XP_011514442.1 | |
| XM_011516141.2 | 1 | 203 | XP_011514443.1 | |
| AK126174.1 | 1 | 203 | BAC86473.1 | |
| BC104963.1 | 1 | 163 | AAI04964.1 | |
| BC104965.1 | 1 | 163 | AAI04966.1 | |
| BC110858.1 | 1 | 150 | AAI10859.1 | |
| DB128604.1 | 1 | 198 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526825 | Chr.7:139442337 - 139483870 on Build GRCh38 | Loss |
|
| esv2735212 | Chr.7:138060002 - 140415947 on Build GRCh38 | Deletion |
|
| nsv526644 | Chr.7:139478717 - 139483870 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map