Assay Details
Target Gene Details
Entrez Gene ID: | 64222 |
Gene Name: | torsin family 3 member A |
Gene Aliases: |
ADIR, ADIR2 |
Location: |
Chr.1:179081977-179095996 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TOR3A | NM_022371.3 | 6 | 1199 | NP_071766.2 |
| AJ299403.1 | 6 | 1088 | CAC13973.1 | |
| AJ299441.1 | CAC14461.1 | |||
| AK025998.1 | 6 | 1078 | BAB15312.1 | |
| AK130132.1 | 4 | 518 | ||
| AK299966.1 | 5 | 796 | ||
| AK316530.1 | 5 | 808 | ||
| BC001085.1 | 6 | 1097 | AAH01085.1 | |
| BC007571.1 | 6 | 1088 | AAH07571.1 | |
| BC011746.1 | 6 | 1084 | AAH11746.1 | |
| BC018292.1 | 6 | 1089 | AAH18292.1 | |
| BC088368.1 | 5 | 826 | AAH88368.1 | |
| CR457354.1 | 6 | 1047 | CAG33635.1 |
More Information
Set Membership: |
|

Genomic Map