Assay Details
Target Gene Details
Entrez Gene ID: | 63908 |
Gene Name: | NSF attachment protein beta |
Gene Aliases: |
SNAP-BETA, SNAPB |
Location: |
Chr.20:23374519-23421519 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NAPB | NM_001283018.1 | NP_001269947.1 | ||
| NM_001283020.1 | NP_001269949.1 | |||
| NM_001283026.1 | NP_001269955.1 | |||
| NM_022080.2 | NP_071363.1 | |||
| NR_104266.1 | ||||
| XM_011529313.1 | XP_011527615.1 | |||
| XM_017028008.1 | XP_016883497.1 | |||
| AK022817.1 | ||||
| AK124876.1 | ||||
| AK125811.1 | ||||
| AK126340.1 | ||||
| AK294703.1 | ||||
| AK295791.1 | ||||
| AK296385.1 | ||||
| AK307709.1 | ||||
| AL834445.1 | CAD39105.1 | |||
| BC047426.1 | AAH47426.1 | |||
| BC060840.1 | AAH60840.1 | |||
| BX648330.1 |
More Information
Set Membership: |
|

Genomic Map