Assay Details
Target Gene Details
Entrez Gene ID: | 284076 |
Gene Name: | tubulin tyrosine ligase like 6 |
Gene Aliases: |
TTL.6 |
Location: |
Chr.17:48762231-48817253 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTLL6 | NM_001130918.1 | 16 | 3025 | NP_001124390.1 |
| NM_173623.3 | 9 | 2113 | NP_775894.2 | |
| XM_017024491.1 | 16 | 3168 | XP_016879980.1 | |
| XM_017024492.1 | 15 | 3281 | XP_016879981.1 | |
| XM_017024493.1 | 17 | 3235 | XP_016879982.1 | |
| XM_017024494.1 | 16 | 3170 | XP_016879983.1 | |
| XM_017024495.1 | 15 | 3898 | XP_016879984.1 | |
| XM_017024496.1 | 11 | 2308 | XP_016879985.1 | |
| XM_017024497.1 | 14 | 2954 | XP_016879986.1 | |
| AK093127.1 | 8 | 1977 | BAC04066.1 | |
| AK302952.1 | 16 | 2971 | ||
| AK310321.1 | 7 | 1509 | ||
| AL834151.1 | 14 | 2955 | CAH56366.1 | |
| BC033563.1 | 9 | 2114 | AAH33563.1 |
More Information
Set Membership: |
|

Genomic Map