Assay Details
Target Gene Details
Entrez Gene ID: | 1741 |
Gene Name: | discs large MAGUK scaffold protein 3 |
Gene Aliases: |
MRX, MRX90, NEDLG, PPP1R82, SAP102, XLMR |
Location: |
Chr.X:70444850-70505490 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DLG3 | NM_021120.3 | NP_066943.2 | ||
| XM_006724625.2 | XP_006724688.1 | |||
| XM_006724626.2 | XP_006724689.1 | |||
| XM_011530883.1 | XP_011529185.1 | |||
| AB209328.1 | BAD92565.1 | |||
| AK315041.1 | ||||
| BC093864.1 | AAH93864.1 | |||
| BC093866.1 | AAH93866.1 | |||
| U49089.1 | AAB61453.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33082 | Chr.X:70420685 - 71888183 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map