Assay Details
Target Gene Details
Entrez Gene ID: | 9493 |
Gene Name: | kinesin family member 23 |
Gene Aliases: |
CHO1, KNSL5, MKLP-1, MKLP1 |
Location: |
Chr.15:69414246-69448427 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KIF23 | NM_001281301.1 | NP_001268230.1 | ||
| NM_004856.6 | NP_004847.2 | |||
| NM_138555.3 | NP_612565.1 | |||
| XM_005254796.2 | XP_005254853.1 | |||
| XM_005254797.2 | XP_005254854.1 | |||
| XM_005254798.2 | XP_005254855.1 | |||
| XM_005254799.2 | XP_005254856.1 | |||
| XM_011522238.1 | XP_011520540.1 | |||
| XM_011522239.2 | XP_011520541.1 | |||
| XM_017022733.1 | XP_016878222.1 | |||
| AK308322.1 | ||||
| AK310402.1 | ||||
| AU133373.1 | ||||
| BC017705.2 | AAH17705.2 | |||
| BC051826.1 | AAH51826.1 | |||
| DB445953.1 | ||||
| DR000298.1 | ||||
| X67155.2 | CAA47628.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2760037 | Chr.15:69342413 - 69499860 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map