Assay Details
Target Gene Details
Entrez Gene ID: | 3707 |
Gene Name: | inositol-trisphosphate 3-kinase B |
Gene Aliases: |
IP3-3KB, IP3K, IP3K-B, IP3KB, PIG37 |
Location: |
Chr.1:226631690-226739327 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ITPKB | NM_002221.3 | 8 | 3906 | NP_002212.3 |
| AJ242780.1 | 7 | 3623 | CAC40650.1 | |
| X57206.1 | 7 | 2245 | CAA40491.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470779 | Chr.1:226590731 - 226645603 on Build GRCh38 | Loss |
|
| nsv522446 | Chr.1:226597829 - 226736237 on Build GRCh38 | Loss |
|
| nsv4755 | Chr.1:226608634 - 226643907 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map