Assay Details
Target Gene Details
Entrez Gene ID: | 80173 |
Gene Name: | intraflagellar transport 74 |
Gene Aliases: |
CCDC2, CMG-1, CMG1 |
Location: |
Chr.9:26947039-27066129 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IFT74 | NM_001099222.1 | NP_001092692.1 | ||
| NM_001099223.1 | NP_001092693.1 | |||
| NM_001099224.1 | NP_001092694.1 | |||
| NM_025103.2 | NP_079379.2 | |||
| XM_011518035.2 | XP_011516337.1 | |||
| AK023707.1 | BAB14650.1 | |||
| AK225570.1 | ||||
| AY040325.1 | AAK77221.1 | |||
| BC023634.2 | ||||
| BC107742.1 | AAI07743.1 | |||
| BC113524.1 |
Target Gene Details
Entrez Gene ID: | 64922 |
Gene Name: | leucine rich repeat containing 19 |
Gene Aliases: |
- |
Location: |
Chr.9:26993136-27005693 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRRC19 | NM_022901.2 | 5 | 1382 | NP_075052.1 |
| AK024955.1 | 5 | 1361 | BAB15042.1 | |
| AK025070.1 | 1 | 262 | ||
| AK222987.1 | 5 | 1354 | BAD96707.1 | |
| BC126156.1 | 5 | 1341 | ||
| BC136298.1 | 5 | 1341 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv517269 | Chr.9:26945978 - 27137244 on Build GRCh38 | Gain |
|
| nsv528614 | Chr.9:26893664 - 27074235 on Build GRCh38 | Gain |
|
| nsv1015259 | Chr.9:26347305 - 27643043 on Build GRCh38 | Gain |
|
| esv3891647 | Chr.9:26743024 - 27123262 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map