Assay Details
Target Gene Details
Entrez Gene ID: | 835 |
Gene Name: | caspase 2 |
Gene Aliases: |
CASP-2, ICH1, NEDD-2, NEDD2, PPP1R57 |
Location: |
Chr.7:143288215-143307696 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CASP2 | NM_001224.4 | NP_001215.1 | ||
| NM_032982.3 | NP_116764.2 | |||
| NM_032983.3 | NP_116765.2 | |||
| AB209640.1 | BAD92877.1 | |||
| AF314174.1 | AAK00299.1 | |||
| AF314175.1 | AAK00300.1 | |||
| AK291274.1 | ||||
| AK303391.1 | ||||
| BC002427.2 | AAH02427.2 | |||
| BG397039.1 | ||||
| BT007240.1 | AAP35904.1 | |||
| CR541748.1 | CAG46548.1 | |||
| KU177957.1 | ||||
| KU177958.1 | ||||
| U13021.1 | AAA58959.1 | |||
| U13022.1 | AAA58960.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831169 | Chr.7:143188183 - 143348025 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map