Assay Details
Target Gene Details
Entrez Gene ID: | 4776 |
Gene Name: | nuclear factor of activated T-cells 4 |
Gene Aliases: |
NF-AT3, NF-ATC4, NFAT3 |
Location: |
Chr.14:24366911-24379604 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NFATC4 | NM_001136022.2 | NP_001129494.1 | ||
| NM_001198965.1 | 1 | 262 | NP_001185894.1 | |
| NM_001198967.2 | NP_001185896.1 | |||
| NM_001320043.1 | NP_001306972.1 | |||
| NM_004554.4 | 1 | 262 | NP_004545.2 | |
| AK293185.1 | 1 | 138 | ||
| AK293873.1 | ||||
| AK309339.1 | ||||
| AK309907.1 | ||||
| AK310445.1 | ||||
| AK311078.1 | 1 | 96 | ||
| AK311425.1 | ||||
| BC053855.1 | 1 | 78 | AAH53855.1 | |
| DC318153.1 | 1 | 262 | ||
| DC403050.1 | ||||
| EU887632.1 | ||||
| EU887633.1 | ||||
| EU887634.1 | ||||
| EU887635.1 | ||||
| EU887636.1 | ||||
| EU887637.1 | ||||
| EU887638.1 | ||||
| EU887639.1 | ||||
| EU887640.1 | ||||
| EU887641.1 | ||||
| EU887642.1 | ||||
| EU887643.1 | ||||
| L41066.1 | 1 | 82 | AAA79175.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952819 | Chr.14:24367695 - 24377694 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map