Assay Details
Target Gene Details
Entrez Gene ID: | 6281 |
Gene Name: | S100 calcium binding protein A10 |
Gene Aliases: |
42C, ANX2L, ANX2LG, CAL1L, CLP11, Ca[1], GP11, P11, p10 |
Location: |
Chr.1:151982910-151994238 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| S100A10 | NM_002966.2 | NP_002957.1 | ||
| BI827113.1 | 1 | 182 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1007097 | Chr.1:151860320 - 152255501 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map