Assay Details
Target Gene Details
Entrez Gene ID: | 963 |
Gene Name: | CD53 molecule |
Gene Aliases: |
MOX44, TSPAN25 |
Location: |
Chr.1:110871199-110899936 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CD53 | NM_000560.3 | NP_000551.1 | ||
| NM_001040033.1 | NP_001035122.1 | |||
| NM_001320638.1 | NP_001307567.1 | |||
| AK298723.1 | ||||
| AK313582.1 | ||||
| BC040693.1 | AAH40693.1 | |||
| BG536920.1 | ||||
| BT019643.1 | AAV38449.1 | |||
| M37033.1 | AAA35663.1 | |||
| M60871.1 | AAA51951.1 |
Target Gene Details
Entrez Gene ID: | 55791 |
Gene Name: | ligand dependent nuclear receptor interacting factor 1 |
Gene Aliases: |
C1orf103, RIF1 |
Location: |
Chr.1:110877252-110964203 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRIF1 | XM_017001769.1 | XP_016857258.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831037 | Chr.1:110770456 - 110911256 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map