Assay Details
Target Gene Details
Entrez Gene ID: | 54545 |
Gene Name: | myotubularin related protein 12 |
Gene Aliases: |
3-PAP, PIP3AP |
Location: |
Chr.5:32227007-32313008 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MTMR12 | NM_001040446.2 | 16 | 3849 | NP_001035536.1 |
| NM_001294343.1 | 15 | 3687 | NP_001281272.1 | |
| NM_001294344.1 | 14 | 3519 | NP_001281273.1 | |
| XM_011514057.1 | 18 | 3969 | XP_011512359.1 | |
| XM_011514058.2 | 17 | 3909 | XP_011512360.1 | |
| XM_011514059.1 | 17 | 3801 | XP_011512361.1 | |
| XM_011514060.1 | 15 | 3579 | XP_011512362.1 | |
| XM_017009589.1 | 15 | 3645 | XP_016865078.1 | |
| XM_017009590.1 | 19 | 3907 | XP_016865079.1 | |
| XM_017009591.1 | 17 | 3918 | XP_016865080.1 | |
| XM_017009592.1 | 15 | 3613 | XP_016865081.1 | |
| XM_017009593.1 | 15 | 3740 | XP_016865082.1 | |
| XM_017009594.1 | 16 | 3801 | XP_016865083.1 | |
| XM_017009595.1 | 14 | 3683 | XP_016865084.1 | |
| XM_017009596.1 | 15 | 3569 | XP_016865085.1 | |
| AB051469.1 | 17 | 3697 | ||
| AK000967.1 | ||||
| AL833231.1 | 6 | 2622 | CAH10604.1 | |
| BC057393.1 | 15 | 3617 | AAH57393.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1021781 | Chr.5:32102613 - 32412387 on Build GRCh38 | Gain |
|
| dgv203e55 | Chr.5:32114037 - 32260077 on Build GRCh38 | Gain |
|
| esv3604580 | Chr.5:31525629 - 32360060 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs76497847] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map