Assay Details
Target Gene Details
Entrez Gene ID: | 94120 |
Gene Name: | synaptotagmin like 3 |
Gene Aliases: |
SLP3 |
Location: |
Chr.6:158644881-158766298 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 5 - Exon 6 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv5562 | Chr.6:158655666 - 158690001 on Build GRCh38 | Insertion |
|
| esv3611400 | Chr.6:158655561 - 158687258 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map