Assay Details
Target Gene Details
Entrez Gene ID: | 10505 |
Gene Name: | ssemaphorin 4F |
Gene Aliases: |
M-SEMA, PRO2353, S4F, SEMAM, SEMAW, m-Sema-M |
Location: |
Chr.2:74654228-74699013 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEMA4F | NM_001271661.1 | 8 | 1226 | NP_001258590.1 |
| NM_001271662.1 | 11 | 1592 | NP_001258591.1 | |
| NM_004263.4 | 12 | 1691 | NP_004254.2 | |
| NR_073398.1 | 10 | 1445 | ||
| NR_073399.1 | 10 | 1498 | ||
| XM_011532475.2 | 12 | 1674 | XP_011530777.1 | |
| XM_011532476.2 | 12 | 1674 | XP_011530778.1 | |
| XM_011532477.2 | 12 | 1674 | XP_011530779.1 | |
| XM_017003172.1 | 11 | 1512 | XP_016858661.1 | |
| XM_017003173.1 | 10 | 1218 | XP_016858662.1 | |
| XM_017003174.1 | 7 | 1073 | XP_016858663.1 | |
| AB021292.1 | 10 | 1364 | ||
| AB022317.1 | 8 | 1092 | BAA75631.1 | |
| AF053369.1 | 4 | 506 | AAF80660.1 | |
| AK075384.1 | 12 | 1644 | BAC11584.1 | |
| AK095587.1 | ||||
| AK291023.1 | 8 | 1174 | ||
| AK312728.1 | 10 | 1421 | ||
| AL136552.1 | 12 | 1653 | CAB66487.1 | |
| BC018361.1 | 8 | 1165 | AAH18361.1 | |
| BC038411.1 | 12 | 1592 | AAH38411.1 | |
| KU178531.1 | 12 | 1542 | ||
| KU178532.1 | 10 | 1270 | ||
| KU178533.1 | 9 | 1176 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3591284 | Chr.2:74673795 - 74824958 on Build GRCh38 | Gain |
|
| nsv1003960 | Chr.2:74630228 - 74769504 on Build GRCh38 | Gain |
|
| nsv1001495 | Chr.2:74598036 - 75093218 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map