Assay Details
Target Gene Details
Entrez Gene ID: | 2266 |
Gene Name: | fibrinogen gamma chain |
Gene Aliases: |
- |
Location: |
Chr.4:154604134-154612808 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FGG | NM_000509.5 | 7 | 999 | NP_000500.2 |
| NM_021870.2 | 7 | 941 | NP_068656.2 | |
| AK289422.1 | 7 | 844 | ||
| AK290824.1 | 7 | 890 | ||
| AL531173.3 | 3 | 790 | ||
| BC007044.1 | 7 | 846 | AAH07044.1 | |
| BC021674.1 | 7 | 848 | AAH21674.1 | |
| BT007081.1 | 7 | 800 | AAP35744.1 | |
| BX382942.2 | ||||
| BX538098.1 | 6 | 688 | CAD98014.1 | |
| X00086.1 | CAA24944.1 | |||
| X51473.1 | 5 | 578 | CAA35837.1 |
More Information
Set Membership: |
|

Genomic Map