Assay Details
Target Gene Details
Entrez Gene ID: | 57592 |
Gene Name: | zinc finger protein 687 |
Gene Aliases: |
PDB6 |
Location: |
Chr.1:151281555-151291905 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF687 | NM_001304763.1 | 10 | 4918 | NP_001291692.1 |
| NM_001304764.1 | 9 | 4119 | NP_001291693.1 | |
| NM_020832.2 | 9 | 4049 | NP_065883.1 | |
| XM_005245366.3 | 9 | 4487 | XP_005245423.1 | |
| XM_011509811.2 | 8 | 3919 | XP_011508113.1 | |
| XM_011509812.2 | 10 | 4366 | XP_011508114.1 | |
| XM_011509813.2 | 9 | 4250 | XP_011508115.1 | |
| AB037862.1 | 10 | 4918 | ||
| AK023105.1 | 8 | 2817 | BAB14406.1 | |
| BC032463.1 | 9 | 4046 | AAH32463.1 | |
| CR749307.1 | 9 | 4120 | CAH18162.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522840 | Chr.1:150961247 - 151375270 on Build GRCh38 | Gain |
|
| esv34461 | Chr.1:151268066 - 151439159 on Build GRCh38 | Gain |
|
| dgv2n68 | Chr.1:151136983 - 151309302 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map