Assay Details
Target Gene Details
Entrez Gene ID: | 2696 |
Gene Name: | gastric inhibitory polypeptide receptor |
Gene Aliases: |
PGQTL2 |
Location: |
Chr.19:45668187-45683724 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GIPR | NM_000164.3 | NP_000155.1 | ||
| NM_001308418.1 | NP_001295347.1 | |||
| XM_011526709.2 | XP_011525011.1 | |||
| XM_011526710.2 | XP_011525012.1 | |||
| XM_011526711.2 | XP_011525013.1 | |||
| XM_011526713.2 | XP_011525015.1 | |||
| XM_011526714.2 | XP_011525016.1 | |||
| XM_011526715.2 | XP_011525017.1 | |||
| XM_011526716.2 | XP_011525018.1 | |||
| XM_017026584.1 | XP_016882073.1 | |||
| XM_017026585.1 | XP_016882074.1 | |||
| XM_017026586.1 | XP_016882075.1 | |||
| XM_017026587.1 | XP_016882076.1 | |||
| BC093723.1 | AAH93723.1 | |||
| BC095494.1 | AAH95494.1 | |||
| BC101673.1 | AAI01674.1 | |||
| BC143300.1 | ||||
| S79852.1 | AAB35419.2 | |||
| U39231.1 | AAA84418.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3644509 | Chr.19:45677013 - 45696301 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map