Assay Details
Target Gene Details
Entrez Gene ID: | 219873 |
Gene Name: | olfactory receptor family 10 subfamily S member 1 |
Gene Aliases: |
OR11-279 |
Location: |
Chr.11:123976696-123977691 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR10S1 | NM_001004474.1 | 1 | 212 | NP_001004474.1 |
| BC137037.1 | 1 | 302 | ||
| BC137038.1 | 1 | 302 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2761716 | Chr.11:123964222 - 123986104 on Build GRCh38 | Loss |
|
| dgv82n21 | Chr.11:123901052 - 124289279 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map