Assay Details
Target Gene Details
Entrez Gene ID: | 253782 |
Gene Name: | ceramide synthase 6 |
Gene Aliases: |
CERS5, LASS6 |
Location: |
Chr.2:168456249-168775134 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CERS6 | NM_001256126.1 | 1 | 298 | NP_001243055.1 |
| NM_203463.2 | 1 | 298 | NP_982288.1 | |
| AI521447.1 | 1 | 222 | ||
| AK172775.1 | 1 | 298 | BAD18757.1 | |
| BC109284.1 | 1 | 148 | AAI09285.1 | |
| BC109285.1 | 1 | 148 | AAI09286.1 | |
| BX393696.2 | 1 | 228 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834450 | Chr.2:168334862 - 168525126 on Build GRCh38 | Gain |
|
| nsv428405 | Chr.2:168301712 - 168627188 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map