Assay Details
Target Gene Details
Entrez Gene ID: | 374407 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member B13 |
Gene Aliases: |
RSPH16A, TSARG5, TSARG6 |
Location: |
Chr.11:73950319-73970287 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJB13 | NM_153614.3 | 3 | 966 | NP_705842.2 |
| XM_005273984.3 | XP_005274041.1 | |||
| XM_011545004.2 | 5 | 1217 | XP_011543306.1 | |
| XM_011545005.2 | XP_011543307.1 | |||
| XM_011545007.2 | 3 | 742 | XP_011543309.1 | |
| XM_011545009.2 | 5 | 891 | XP_011543311.1 | |
| XM_011545013.2 | 5 | 1223 | XP_011543315.1 | |
| XM_011545014.2 | 5 | 1223 | XP_011543316.1 | |
| XM_017017675.1 | 4 | 487 | XP_016873164.1 | |
| XM_017017676.1 | 5 | 1067 | XP_016873165.1 | |
| XM_017017677.1 | 4 | 936 | XP_016873166.1 | |
| XM_017017678.1 | 3 | 747 | XP_016873167.1 | |
| XM_017017679.1 | 5 | 1223 | XP_016873168.1 | |
| AK128589.1 | 3 | 259 | ||
| AY138810.2 | 3 | 966 | AAN15929.1 | |
| AY325766.1 | 3 | 259 |
More Information
Set Membership: |
|

Genomic Map